The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178020.3 | NP_821139.2 | 2 - 3 | 953 | 94 |
GenBank mRNA | AF074489.1 | - | 1 - 2 | 909 | 94 |
AY048681.1 | - | 2 - 3 | 942 | 94 | |
BC018457.1 | - | 2 - 3 | 953 | 94 |