The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177396.1 | NP_796370.1 | 4 - 5 | 484 | 142 |
GenBank mRNA | AY184375.1 | - | 4 - 5 | 484 | 142 |