The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083927.1 | NP_001077396.1 | 1 - 2 | 1093 | 58 |
NM_001083928.1 | NP_001077397.1 | 1 - 2 | 1093 | 58 | |
NM_009389.2 | NP_033415.2 | 1 - 2 | 1093 | 58 | |
GenBank mRNA | AK122524.1 | - | 1 - 2 | 601 | 58 |
BC056465.1 | - | 1 - 2 | 74 | 58 |