Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mapkapk2
Entrez Gene ID
17164
Gene Name
MAP kinase-activated protein kinase 2
Gene Aliases
AA960234, MAPKAP-K2, MK-2, MK2, Rps6kc1
Chromosome Location
Chr.1: 131053700 - 131097543 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008551.1 NP_032577.1 9 - 10 1031 60
GenBank mRNA AK192111.1 - 2 - 3 149 60
AK198261.1 - 2 - 3 149 60
BC024559.1 - 5 - 6 400 60
BC063064.1 - 9 - 10 1031 60
X76850.1 - 9 - 10 1010 60

View Full Product Details