The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253831.1 | NP_001240760.1 | 12 - 13 | 1235 | 115 |
NM_001253834.1 | NP_001240763.1 | 13 - 14 | 1326 | 115 | |
NM_001253836.1 | NP_001240765.1 | 12 - 13 | 1258 | 115 | |
NM_175025.4 | NP_778190.3 | 13 - 14 | 1441 | 115 | |
GenBank mRNA | AJ551270.1 | - | 11 - 12 | 1023 | 115 |
AK028393.1 | - | 12 - 13 | 1204 | 115 | |
AK159918.1 | - | 13 - 14 | 1392 | 115 | |
AK163049.1 | - | 13 - 14 | 1287 | 115 | |
AK173154.1 | - | 11 - 12 | 1014 | 115 | |
BC043091.1 | - | 13 - 14 | 1407 | 115 | |
BC043706.1 | - | 13 - 14 | 1388 | 114 |