Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mael
Entrez Gene ID
98558
Gene Name
maelstrom homolog (Drosophila)
Gene Aliases
4933405K18Rik, AU019877
Chromosome Location
Chr.1: 166201385 - 166238744 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175296.4 NP_780505.1 7 - 8 780 75
GenBank mRNA AK077103.1 - 7 - 8 777 75
AK165801.1 - 7 - 8 777 75
BC050800.1 - 7 - 8 780 75

View Full Product Details