The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177178.3 | NP_796152.2 | 17 - 18 | 2256 | 131 |
GenBank mRNA | AK036527.1 | - | 17 - 18 | 2278 | 131 |
AK051777.1 | - | 11 - 12 | 1263 | 131 | |
AK079450.1 | - | 17 - 18 | 2256 | 131 | |
AK153728.1 | - | 17 - 18 | 2207 | 131 | |
BC095971.1 | - | 17 - 18 | 2038 | 131 | |
BC120727.1 | - | 17 - 18 | 2228 | 131 | |
BC125421.1 | - | 17 - 18 | 2228 | 131 |