Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl18
Entrez Gene ID
270201
Gene Name
kelch-like 18
Gene Aliases
A930041K15, AW545966
Chromosome Location
Chr.9: 110425926 - 110476700 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC025563.1 - 6 - 7 997 91

View Full Product Details