Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atl1
Entrez Gene ID
73991
Gene Name
atlastin GTPase 1
Gene Aliases
4930435M24Rik, Adfsp, Fsp1, Spg3, Spg3a
Chromosome Location
Chr.12: 69893105 - 69964085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178628.5 NP_848743.1 11 - 12 1441 70
GenBank mRNA AK046919.1 - 10 - 11 1171 70
AK050339.1 - 11 - 12 1278 70
AK083918.1 - - 307 70
AK147311.1 - 11 - 12 4705 70
BC057068.1 - 6 - 7 1002 70

View Full Product Details