Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atl1
Entrez Gene ID
73991
Gene Name
atlastin GTPase 1
Gene Aliases
4930435M24Rik, Adfsp, Fsp1, Spg3, Spg3a
Chromosome Location
Chr.12: 69893105 - 69964085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178628.5 NP_848743.1 12 - 13 1516 84
GenBank mRNA AK046919.1 - 11 - 12 1246 84
AK050339.1 - 12 - 13 1353 84
AK083918.1 - - 382 84
AK140185.1 - 11 - 12 1260 84
AK147311.1 - 12 - 13 4780 84
BC050784.1 - 11 - 12 1245 84
BC057068.1 - 7 - 8 1077 84

View Full Product Details