The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178637.2 | NP_848752.1 | 5 - 6 | 501 | 119 |
GenBank mRNA | AF528194.1 | - | 1 - 2 | 55 | 119 |
AF528195.1 | - | 4 - 5 | 436 | 119 | |
AK146352.1 | - | 5 - 6 | 458 | 119 | |
AK219943.1 | - | 1 - 2 | 94 | 119 | |
BC110675.1 | - | 5 - 6 | 442 | 119 | |
DQ139980.1 | - | 4 - 5 | 436 | 119 |