The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178768.4 | NP_848883.2 | 1 - 2 | 247 | 63 |
GenBank mRNA | AK049048.1 | - | 1 - 2 | 402 | 63 |
AK049938.1 | - | 1 - 2 | 360 | 63 | |
AK085603.1 | - | 1 - 2 | 248 | 63 | |
BC093486.1 | - | 1 - 2 | 390 | 63 | |
BC116440.1 | - | 1 - 2 | 231 | 63 | |
BC116441.1 | - | 1 - 2 | 231 | 63 |