Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc85a
Entrez Gene ID
216613
Gene Name
coiled-coil domain containing 85A
Gene Aliases
-
Chromosome Location
Chr.11: 28385684 - 28584441 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181577.5 NP_853555.2 4 - 5 2197 63
GenBank mRNA AK045568.1 - 4 - 5 2170 63
BC020949.1 - 3 - 4 1160 63

View Full Product Details