The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011029.4 | NP_035159.3 | 7 - 7 | 1579 | 121 |
GenBank mRNA | AK082935.1 | - | 7 - 7 | 1559 | 121 |