Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tceb1
- Entrez Gene ID
-
67923
- Gene Name
- transcription elongation factor B (SIII), polypeptide 1
- Gene Aliases
-
2610043E24Rik, 2610301I15Rik, AA407206, AI987979, AW049146
- Chromosome Location
-
Chr.1: 16641725 - 16657097 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK039746.1 |
- |
4 - 4 |
1354 |
70 |
View Full Product Details