Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Impdh2
Entrez Gene ID
23918
Gene Name
inosine 5'-phosphate dehydrogenase 2
Gene Aliases
IMPD
Chromosome Location
Chr.9: 108560291 - 108565584 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011830.3 NP_035960.2 13 - 14 1560 89
GenBank mRNA AK002676.1 - 13 - 14 1592 89
AK011777.1 - 13 - 14 1587 89
AK143505.1 - 13 - 14 1601 89
AK146362.1 - 13 - 14 1625 89
AK150713.1 - 13 - 14 1596 89
AK151233.1 - 13 - 14 1596 89
AK151706.1 - 13 - 14 1619 89
AK152691.1 - 13 - 14 1593 89
AK159822.1 - 13 - 14 1593 89
AK160569.1 - 13 - 14 1593 89
AK166667.1 - 13 - 14 1595 89
AK169129.1 - 13 - 14 1626 89
AK169788.1 - 13 - 14 1626 89
AK203194.1 - 3 - 4 307 89
BC010314.1 - 13 - 14 1560 89
BC052671.1 - 13 - 14 1581 89
M33934.1 - 13 - 14 1593 89

View Full Product Details