The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175423.3 | NP_780632.1 | 1 - 2 | 489 | 150 |
GenBank mRNA | AB271214.1 | - | 1 - 2 | 305 | 150 |
AK052573.1 | - | 1 - 2 | 490 | 150 | |
AK085755.1 | - | 1 - 2 | 240 | 150 | |
AM712355.1 | - | 1 - 2 | 305 | 150 | |
BC023149.1 | - | 1 - 2 | 261 | 150 |