The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173184.1 | NP_775276.1 | 1 - 2 | 213 | 67 |
GenBank mRNA | AB076565.1 | - | 1 - 2 | 213 | 67 |