The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039079.2 | NP_001034168.1 | 14 - 15 | 1699 | 159 |
NM_008860.3 | NP_032886.2 | 17 - 18 | 1899 | 159 | |
GenBank mRNA | AB110830.1 | - | 14 - 15 | 1659 | 159 |
AK042562.1 | - | 14 - 15 | 1657 | 159 | |
AK049959.1 | - | 14 - 15 | 1683 | 159 | |
AK147300.1 | - | 14 - 15 | 1517 | 159 | |
BC057694.1 | - | 9 - 10 | 866 | 159 | |
BC072590.1 | - | 14 - 15 | 1699 | 159 | |
BC139761.1 | - | 17 - 18 | 1716 | 159 | |
M94632.1 | - | 17 - 18 | 1716 | 159 |