The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008944.2 | NP_032970.2 | 4 - 5 | 405 | 136 |
GenBank mRNA | AF099733.1 | - | 4 - 5 | 372 | 136 |
AK012119.1 | - | 4 - 5 | 387 | 136 | |
AK012990.1 | - | 4 - 5 | 394 | 136 | |
AK035578.1 | - | - | 394 | 136 | |
AK136115.1 | - | 4 - 5 | 407 | 136 | |
AK151883.1 | - | 4 - 5 | 396 | 136 | |
AK152862.1 | - | 4 - 5 | 396 | 136 | |
AK154011.1 | - | 4 - 5 | 389 | 136 | |
AK167752.1 | - | 4 - 5 | 396 | 136 | |
BC026768.1 | - | 4 - 5 | 378 | 136 | |
CJ052982.1 | - | 4 - 5 | 407 | 136 | |
X70303.1 | - | 4 - 5 | 449 | 136 |