The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011731.3 | NP_035861.2 | 2 - 3 | 595 | 158 |
GenBank mRNA | AF075261.1 | - | 2 - 3 | 419 | 158 |
AK076806.1 | - | 3 - 4 | 329 | 158 | |
AK143893.1 | - | 2 - 3 | 565 | 158 | |
AK144038.1 | - | 2 - 3 | 596 | 158 | |
AK163340.1 | - | 3 - 4 | 484 | 158 | |
AK164706.1 | - | 2 - 3 | 440 | 158 | |
AK164713.1 | - | 2 - 3 | 430 | 158 | |
BC013484.1 | - | 2 - 3 | 506 | 158 |