The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177621.1 | NP_001171092.1 | 18 - 18 | 3387 | 92 |
NM_001177622.1 | NP_001171093.1 | 18 - 18 | 3169 | 92 | |
NM_177870.5 | NP_808538.1 | 17 - 17 | 3276 | 92 | |
GenBank mRNA | AK088682.1 | - | 18 - 18 | 3075 | 92 |
AK158896.1 | - | 18 - 18 | 3388 | 92 | |
AK161633.1 | - | 17 - 17 | 3200 | 92 | |
BC065061.1 | - | 18 - 18 | 3168 | 92 | |
BC085132.1 | - | 17 - 17 | 2972 | 92 |