Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aprt
Entrez Gene ID
11821
Gene Name
adenine phosphoribosyl transferase
Gene Aliases
C85684
Chromosome Location
Chr.8: 122574637 - 122576907 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009698.2 NP_033828.2 5 - 5 692 135
GenBank mRNA AK002350.1 - 5 - 5 645 135
AK153201.1 - 5 - 5 657 135
BC005667.1 - 5 - 5 630 135

View Full Product Details