The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009846.2 | NP_033976.1 | 2 - 2 | 780 | 142 |
GenBank mRNA | AK014279.1 | - | 2 - 2 | 780 | 142 |
AK133869.1 | - | 2 - 2 | 779 | 142 | |
AK168111.1 | - | 2 - 2 | 779 | 142 | |
AK169613.1 | - | 2 - 2 | 777 | 142 | |
BC023197.1 | - | 1 - 1 | 504 | 142 | |
BC075622.1 | - | 2 - 2 | 760 | 142 | |
M58661.1 | - | 2 - 2 | 768 | 142 | |
X53825.1 | - | 2 - 2 | 718 | 142 |