Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Immp1l
Entrez Gene ID
66541
Gene Name
IMP1 inner mitochondrial membrane peptidase-like (S. cerevisiae)
Gene Aliases
1500034J20Rik, 2610528O17Rik, AU042386
Chromosome Location
Chr.2: 105904638 - 105965558 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK012181.1 - - 839 102

View Full Product Details