The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011781.2 | NP_035911.2 | 2 - 2 | 1040 | 123 |
GenBank mRNA | AF167403.1 | - | - | 1040 | 123 |
AK016581.1 | - | 2 - 2 | 1038 | 123 |