Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Xlr4b
Entrez Gene ID
27083
Gene Name
X-linked lymphocyte-regulated 4B
Gene Aliases
Xlr4
Chromosome Location
Chr.X: 73214323 - 73222453 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293676.1 NP_001280605.1 6 - 6 579 125
NM_001293677.1 NP_001280606.1 6 - 6 532 125
NM_001293678.1 NP_001280607.1 6 - 6 556 125
NM_001293679.1 NP_001280608.1 6 - 6 568 125
NM_001293680.1 NP_001280609.1 6 - 6 592 125
NM_021365.3 NP_067340.2 6 - 6 696 125
GenBank mRNA AL136330.1 - 6 - 6 664 125
BC025576.1 - 6 - 6 532 125
BC034848.1 - 6 - 6 1120 125
BC138975.1 - 6 - 6 438 125
BC138979.1 - 6 - 6 462 125
BC145481.1 - 6 - 6 462 125
BC145482.1 - 6 - 6 474 125
BC145483.1 - 6 - 6 498 125
BE627633.1 - 6 - 6 495 125

Gene Symbol
Xlr4a
Entrez Gene ID
434794
Gene Name
X-linked lymphocyte-regulated 4A
Gene Aliases
-
Chromosome Location
Chr.X: 73074345 - 73082507 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081642.1 NP_001075111.1 6 - 6 568 125
GenBank mRNA BC098242.1 - 6 - 6 548 125

Gene Symbol
Xlr4c
Entrez Gene ID
72891
Gene Name
X-linked lymphocyte-regulated 4C
Gene Aliases
2900036K24Rik
Chromosome Location
Chr.X: 73234072 - 73243130 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183094.3 NP_898917.2 6 - 6 598 125
GenBank mRNA AK166871.1 - 6 - 6 599 125

View Full Product Details