The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007696.2 | NP_031722.1 | 9 - 10 | 1031 | 70 |
GenBank mRNA | AY521455.1 | - | 9 - 10 | 1017 | 70 |
BC137995.1 | - | 9 - 10 | 1122 | 70 | |
D32137.1 | - | 9 - 10 | 1031 | 70 |