Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plxna2
Entrez Gene ID
18845
Gene Name
plexin A2
Gene Aliases
2810428A13Rik, AA589422, AW457381, OCT, PlexA2, Plxn2, mKIAA0463
Chromosome Location
Chr.1: 194619829 - 194816869 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008882.2 NP_032908.2 28 - 29 5699 70
GenBank mRNA AK122289.1 - 27 - 28 3963 70
BC024509.1 - 3 - 4 359 70
BC051045.1 - 2 - 3 485 70
BC056475.1 - 8 - 9 1150 70
BC068155.1 - 28 - 29 5481 70
D86949.1 - 28 - 29 5605 70

View Full Product Details