The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009713.4 | NP_033843.2 | 5 - 6 | 2051 | 67 |
GenBank mRNA | AK004540.1 | - | 4 - 5 | 1270 | 67 |
AK029854.1 | - | 3 - 4 | 1312 | 67 | |
AK132501.1 | - | 4 - 5 | 1282 | 67 | |
AK165921.1 | - | 4 - 5 | 1236 | 67 | |
BC011284.1 | - | 4 - 5 | 1233 | 67 | |
BC098075.1 | - | 5 - 6 | 2063 | 67 | |
X73230.1 | - | 4 - 5 | 1484 | 67 |