Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hp1bp3
Entrez Gene ID
15441
Gene Name
heterochromatin protein 1, binding protein 3
Gene Aliases
Hp1bp74
Chromosome Location
Chr.4: 138216582 - 138244683 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122896.2 NP_001116368.1 8 - 9 1014 92
NM_001122897.2 NP_001116369.1 9 - 10 1251 92
NM_001285478.1 NP_001272407.1 9 - 10 1271 92
NM_001285479.1 NP_001272408.1 9 - 10 1209 92
NM_001285480.1 NP_001272409.1 8 - 9 975 92
NM_001285481.1 NP_001272410.1 9 - 10 1173 92
NM_010470.3 NP_034600.2 9 - 10 1212 92
GenBank mRNA AK004938.1 - 9 - 10 1189 92
AK028181.1 - 9 - 10 1187 92
AK043260.1 - 8 - 9 988 92
AK050234.1 - 9 - 10 1191 92
AK075725.1 - 8 - 9 952 92
AK150784.1 - 9 - 10 1255 92
AK161041.1 - 8 - 9 982 92
AK166161.1 - 9 - 10 1159 92
AK169740.1 - 9 - 10 1118 92
AK205238.1 - 1 - 2 45 92
BC004053.1 - 5 - 6 492 92
BC020024.1 - 9 - 10 1162 92

View Full Product Details