Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhx8
Entrez Gene ID
16875
Gene Name
LIM homeobox protein 8
Gene Aliases
L3, Lhx7
Chromosome Location
Chr.3: 154306288 - 154330560 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010713.2 NP_034843.2 7 - 8 1078 60
GenBank mRNA AJ000338.1 - 7 - 8 1178 60
AK160121.1 - 7 - 8 1079 60
AK165733.1 - 7 - 8 1072 60
BC125281.1 - 7 - 8 987 60
BC125283.1 - 7 - 8 987 60
BC144768.1 - 7 - 8 987 60
D49658.1 - 7 - 8 1047 60

View Full Product Details