Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Theg
Entrez Gene ID
21830
Gene Name
testicular haploid expressed gene
Gene Aliases
-
Chromosome Location
Chr.10: 79576477 - 79587136 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011583.3 NP_035713.1 3 - 4 547 68
GenBank mRNA AB033128.1 - 3 - 4 529 68
AJ011834.1 - 3 - 4 732 68

View Full Product Details