Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Theg
Entrez Gene ID
21830
Gene Name
testicular haploid expressed gene
Gene Aliases
-
Chromosome Location
Chr.10: 79576477 - 79587136 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011583.3 NP_035713.1 4 - 5 613 64
NM_199226.1 NP_954696.1 3 - 4 541 64
GenBank mRNA AB033128.1 - 4 - 5 595 64
AJ011834.1 - 4 - 5 798 64
BC050970.1 - 3 - 4 542 64
BC061243.1 - 3 - 4 487 64

View Full Product Details