Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sema4b
Entrez Gene ID
20352
Gene Name
sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B
Gene Aliases
SemC, Semac, mKIAA1745
Chromosome Location
Chr.7: 80186841 - 80226527 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013659.4 NP_038687.2 8 - 9 1101 78
GenBank mRNA AK028896.1 - 8 - 9 1064 78
AK030331.1 - 7 - 8 1101 78
AK134973.1 - 7 - 8 1032 78
AK145570.1 - 8 - 9 1102 78
AK154969.1 - 7 - 8 882 78
AK173249.1 - 8 - 9 1049 78
DQ073331.1 - 7 - 8 857 78
X85992.1 - 6 - 7 693 78

View Full Product Details