Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nrip2
Entrez Gene ID
60345
Gene Name
nuclear receptor interacting protein 2
Gene Aliases
AW491344, NIX1
Chromosome Location
Chr.6: 128397625 - 128409784 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162858.1 NP_001156330.1 3 - 4 765 70
NM_021717.2 NP_068363.2 3 - 4 765 70
GenBank mRNA AJ278170.1 - 3 - 4 788 70
AK156526.1 - 3 - 4 638 70
BC094230.1 - 2 - 3 1888 70
BC115523.1 - 3 - 4 515 70

View Full Product Details