The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026452.2 | NP_080728.1 | 8 - 9 | 905 | 63 |
GenBank mRNA | AK004527.1 | - | 4 - 5 | 330 | 63 |
AK009179.1 | - | 8 - 9 | 917 | 63 | |
AK154343.1 | - | 8 - 9 | 930 | 63 | |
AK158979.1 | - | 8 - 9 | 924 | 63 | |
BC036386.1 | - | 8 - 9 | 905 | 63 |