The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027373.2 | NP_081649.1 | 12 - 13 | 1758 | 62 |
GenBank mRNA | AK129483.1 | - | 12 - 13 | 1685 | 62 |
AK147706.1 | - | 12 - 13 | 1650 | 62 | |
AK148000.1 | - | 12 - 13 | 1613 | 62 | |
AK161385.1 | - | 12 - 13 | 1709 | 62 | |
BC050814.1 | - | 12 - 13 | 1758 | 62 |