The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141975.1 | NP_001135447.1 | 6 - 7 | 774 | 73 |
NM_001141976.1 | NP_001135448.1 | 6 - 7 | 790 | 73 | |
NM_001141977.1 | NP_001135449.1 | 6 - 7 | 978 | 73 | |
NM_001141978.1 | NP_001135450.1 | 6 - 7 | 777 | 73 | |
NM_028109.4 | NP_082385.3 | 6 - 7 | 965 | 73 | |
GenBank mRNA | AK011311.2 | - | 3 - 4 | 333 | 73 |
AK090101.1 | - | 6 - 7 | 728 | 73 | |
AK153957.1 | - | 6 - 7 | 712 | 73 | |
AK167030.1 | - | 3 - 4 | 430 | 73 | |
BU612544.1 | - | 2 - 3 | 222 | 73 |