The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277305.1 | NP_001264234.1 | 20 - 21 | 3171 | 71 |
NM_175643.3 | NP_783574.1 | 20 - 21 | 3171 | 71 | |
GenBank mRNA | AK040370.1 | - | 14 - 15 | 1915 | 71 |
AK220464.1 | - | 19 - 20 | 2913 | 71 | |
BC046456.1 | - | 20 - 21 | 3171 | 71 |