Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pik3r5
Entrez Gene ID
320207
Gene Name
phosphoinositide-3-kinase, regulatory subunit 5, p101
Gene Aliases
AV230647, F730038I15Rik
Chromosome Location
Chr.11: 68432125 - 68497849 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177320.2 NP_796294.2 11 - 12 1909 88
GenBank mRNA AK089483.1 - 11 - 12 1914 88
AK154643.1 - 11 - 12 1888 88
AK161050.1 - 2 - 3 817 88
AK171279.1 - 10 - 11 2034 88
AY156924.1 - 10 - 11 1758 88
BC128076.1 - 10 - 11 1758 88
BC128077.1 - 10 - 11 1758 88

View Full Product Details