The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181820.2 | NP_861541.2 | 14 - 15 | 2088 | 72 |
GenBank mRNA | AY236493.1 | - | 14 - 15 | 2051 | 72 |
AY263162.1 | - | 14 - 15 | 2059 | 72 | |
BC022758.1 | - | 6 - 7 | 778 | 72 | |
BC040466.1 | - | 14 - 15 | 2000 | 72 | |
CN719678.1 | - | 3 - 4 | 282 | 72 |