Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxj1
Entrez Gene ID
15223
Gene Name
forkhead box J1
Gene Aliases
FKHL-13, HFH-4, Hfh4
Chromosome Location
Chr.11: 116330704 - 116335516 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008240.3 NP_032266.3 2 - 3 807 98
GenBank mRNA AK140847.1 - 2 - 3 744 98
BC082543.1 - 2 - 3 800 98
BC094569.1 - - 2352 98
L13204.1 - 1 - 2 654 98

View Full Product Details