The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113532.1 | NP_001107004.1 | 6 - 6 | 3103 | 150 |
NM_175394.2 | NP_780603.1 | 6 - 6 | 3151 | 150 | |
GenBank mRNA | AK041341.1 | - | 1 - 1 | 1041 | 150 |
BC046416.1 | - | 6 - 6 | 3030 | 150 | |
BC047338.1 | - | 6 - 6 | 3030 | 150 |