Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spire1
Entrez Gene ID
68166
Gene Name
spire homolog 1 (Drosophila)
Gene Aliases
6030430B19Rik, AI415299, AU022898, AW550622, Spir-1
Chromosome Location
Chr.18: 67488209 - 67610902 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176832.2 NP_789802.1 10 - 11 1187 79
NM_194355.2 NP_919336.1 11 - 12 1459 79
GenBank mRNA BC046486.1 - 4 - 5 542 79
BC058669.1 - 11 - 12 1327 79
BC079633.1 - 1 - 2 35 79
BC094375.1 - 11 - 12 1380 79
BQ178805.1 - 3 - 4 352 79

View Full Product Details