The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009321.2 | NP_033347.1 | 4 - 4 | 414 | 148 |
GenBank mRNA | AK010859.1 | - | 4 - 4 | 431 | 148 |
AK140276.1 | - | 4 - 4 | 438 | 148 | |
AK151218.1 | - | 4 - 4 | 415 | 148 | |
AK168296.1 | - | 4 - 4 | 415 | 148 | |
BC034267.1 | - | 4 - 4 | 405 | 148 | |
BC051475.1 | - | 4 - 4 | 422 | 148 | |
BC055749.1 | - | 4 - 4 | 422 | 148 | |
BC087931.1 | - | 4 - 4 | 437 | 148 | |
U05333.1 | - | 4 - 4 | 413 | 147 |