The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009186.4 | NP_033212.1 | 5 - 6 | 792 | 162 |
GenBank mRNA | AK049573.1 | - | 5 - 6 | 793 | 162 |
AK150007.1 | - | 5 - 6 | 792 | 162 | |
AK152519.1 | - | 5 - 6 | 787 | 162 | |
AK152761.1 | - | 5 - 6 | 793 | 162 | |
AK167387.1 | - | 5 - 6 | 791 | 162 | |
AK167544.1 | - | 5 - 6 | 793 | 162 | |
BC057996.1 | - | 6 - 7 | 968 | 162 | |
BC061177.1 | - | 5 - 6 | 789 | 162 | |
X80232.1 | - | 5 - 6 | 784 | 162 |