The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019426.2 | NP_062299.2 | 4 - 5 | 2014 | 137 |
GenBank mRNA | AK132359.1 | - | 3 - 4 | 634 | 137 |
AK134252.1 | - | 4 - 5 | 2021 | 137 | |
AK167542.1 | - | 5 - 6 | 2060 | 137 | |
BC037617.1 | - | - | 2016 | 137 | |
BC052460.1 | - | 4 - 5 | 2014 | 137 | |
BC060246.1 | - | 4 - 5 | 1997 | 137 |