The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145380.2 | NP_663355.1 | 2 - 3 | 216 | 128 |
GenBank mRNA | AK077480.1 | - | 2 - 3 | 217 | 128 |
AK084738.1 | - | 2 - 3 | 235 | 128 | |
AK146222.1 | - | 2 - 3 | 248 | 128 | |
AK168066.1 | - | 2 - 3 | 233 | 128 | |
BC005598.1 | - | 2 - 3 | 216 | 128 | |
BC103795.1 | - | 2 - 3 | 216 | 128 | |
BC116787.1 | - | 2 - 3 | 213 | 128 | |
BC116789.1 | - | 2 - 3 | 213 | 128 |