Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aida
Entrez Gene ID
108909
Gene Name
axin interactor, dorsalization associated
Gene Aliases
2610208M17Rik, BC004835
Chromosome Location
Chr.1: 183297060 - 183324501 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181732.4 NP_859421.1 6 - 7 914 194
GenBank mRNA AK076031.1 - 6 - 7 566 194
AK081494.1 - 6 - 7 915 194
AK085039.1 - 4 - 5 408 194
AK143716.1 - 6 - 7 454 194
AK149788.1 - 6 - 7 666 194
AK150639.1 - 6 - 7 602 194
AK150702.1 - 6 - 7 663 194
AK151783.1 - 6 - 7 604 194
AK152961.1 - 6 - 7 605 194
AK153448.1 - 6 - 7 602 194
AK158103.1 - 6 - 7 548 194
AK159318.1 - 6 - 7 926 194
AK167520.1 - 6 - 7 631 194
AK172451.1 - 6 - 7 607 194
BC086763.1 - 6 - 7 619 194

View Full Product Details